Merck
  • Genetic immunization: a new monoclonal antibody for the detection of BCL-6 protein in paraffin sections.
Genetic immunization: a new monoclonal antibody for the detection of BCL-6 protein in paraffin sections.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (2005-07-28)
José-Francisco García, Juan-Fernando García, Lorena Maestre, Elena Lucas, Lydia Sánchez-Verde, Silvia Romero-Chala, Miguel-Angel Piris, Giovanna Roncador
ABSTRACT
Genetic immunization can be combined with hybridoma technology to generate high-affinity monoclonal antibodies (MAbs). A new anti-BCL-6 MAb (GI191E/A8) was produced by cloning full-length BCL-6 cDNA into a eukaryotic vector and delivering this into mouse epidermis using a helium gene gun. A comparative study was made of the specificity and the effects of formalin fixation on immunohistochemistry quality of GI191E/A8 and two other anti-BCL-6 MAbs. To evaluate its possible application to differential diagnosis of lymphomas, two tissue microarrays (89 diffuse large B-cell lymphomas and 24 B-cell chronic lymphocytic leukemia cases) were stained with GI191E/A8 and another anti-BCL-6 MAb produced by conventional means. Using GI191E/A8, the detection of BCL-6 protein was significantly increased, and its specificity was independent of formalin-fixation time. Using automatic quantified analysis, the correlation between the two anti-BCL-6 MAbs tested was identical in cases with overexpression or absence of BCL-6. In cases with intermediate BCL-6 protein expression, detection with GI191E/A8 was more sensitive. A significant association of higher BCL-6 expression and longer median overall survival times in diffuse large B-cell lymphomas was found. Using conventionally produced MAbs in the same patient group, the association was not significant.
Sigma-Aldrich
BCL6 (EP278) Rabbit Monoclonal Primary Antibody