Merck
Reduced number of CD1a+ cells in cutaneous B-cell lymphoma.
American journal of clinical pathology (2001-07-13)
M Schmuth, A Sidoroff, B Danner, G Topar, N T Sepp
ABSTRACT
Cutaneous B-cell lymphoma is difficult to distinguish from pseudolymphoma. The histologic pattern and monoclonal restriction (immunohistochemical analysis and molecular biology) are the criteria used for differentiating these entities. CD1a+ dendritic cells have been observed in the infiltrates of T-cell lymphoma, but the presence of these CD1a+ cells has not been compared in B-cell lymphoma and pseudolymphoma. We studied the presence of CD1a+ cells on frozen sections of 23 B-cell lymphomas, 13 pseudolymphomas, and 17 T-cell lymphomas by immunohistochemical analysis. We found abundant CD1a+ dendritic cells in only 1 (4%) of 23 B-cell lymphomas, whereas in 8 (62%) of 13 pseudolymphomas and 17 (100%) of 17 T-cell lymphomas, strong CD1a staining was present. Our study demonstrates a distinct pattern of CD1a staining in the infiltrates of B-cell lymphoma and pseudolymphoma that may be of value in the differential diagnosis of these skin disorders.