- Direct synthesis of tricyclic 5H-pyrido[3,2,1-ij]quinolin-3-one by domino palladium catalyzed reaction.
Direct synthesis of tricyclic 5H-pyrido[3,2,1-ij]quinolin-3-one by domino palladium catalyzed reaction.
Organic & biomolecular chemistry (2006-10-07)
Sébastien Noël, Catherine Pinel, Laurent Djakovitch
PMID17024281
ABSTRACT
Unexpectedly, the palladium catalyzed coupling reaction of acrolein with 8-bromoquinoline gave 5H-pyrido[3,2,1-ij]quinolin-3-one in a single step.
MATERIALS