Merck
  • Reactivity of the bifunctional ambiphilic molecule 8-(dimesitylboryl)quinoline: hydrolysis and coordination to Cu(I), Ag(I) and Pd(II).
Reactivity of the bifunctional ambiphilic molecule 8-(dimesitylboryl)quinoline: hydrolysis and coordination to Cu(I), Ag(I) and Pd(II).
Dalton transactions (Cambridge, England : 2003) (2010-10-23)
Jung-Ho Son, Michael A Pudenz, James D Hoefelmeyer
ABSTRACT
The ambiphilic molecule 8-(dimesitylboryl)quinoline (1) was synthesized by treatment of 8-bromoquinoline or 8-iodoquinoline with n-BuLi followed by dimesitylboronfluoride. Hydrolysis of 1 is unusually rapid compared to bulky triorganoboranes with the sequential loss of mesitylene and formation of mesityl(quinolin-8-yl)borinic acid (2) and 8-quinoline boronic acid dimer (3). Cooperativity within the bifunctional ambiphilic site leads to water activation and protodeboronation of the B-C(Mes) bonds. Blocking of the ambiphilic site of 1 by methylation of the quinoline nitrogen atom leads to an air-stable N-methyl-quinolinium salt. Coordination complexes were formed by reaction of 1 with CuCl, Ag(OTf), and PdCl(2)(PhCN)(2) with coordination of the quinolinyl nitrogen to the metal ion. The Cu(I) and Ag(I) centers are stabilized by η(3)-BC(ipso)C(ortho)π-interaction. The isolated Pd(II) complex is a product of cyclometalation, resulting from elimination of HCl upon deprotonation of the ortho-methyl group of nearby mesityl. The bonding in 7 could be understood as a 16-electron Pd complex that features an anionic η(3)-C(ipso)-C(ortho)-C(benzyl) allylic ligand fragment plus a Pd→B bond, or an η(4)-BC(ipso)-C(ortho)-C(benzyl) boratabutadiene anion fragment.
Sigma-Aldrich
8-Bromoquinoline, 98%