Merck
  • Determination of organic compounds in water using dispersive liquid-liquid microextraction.
Determination of organic compounds in water using dispersive liquid-liquid microextraction.
Journal of chromatography. A (2006-04-01)
Mohammad Rezaee, Yaghoub Assadi, Mohammad-Reza Milani Hosseini, Elham Aghaee, Fardin Ahmadi, Sana Berijani
ABSTRACT
A new microextraction technique termed dispersive liquid-liquid microextraction (DLLME) was developed. DLLME is a very simple and rapid method for extraction and preconcentration of organic compounds from water samples. In this method, the appropriate mixture of extraction solvent (8.0 microL C2Cl4) and disperser solvent (1.00 mL acetone) are injected into the aqueous sample (5.00 mL) by syringe, rapidly. Therefore, cloudy solution is formed. In fact, it is consisted of fine particles of extraction solvent which is dispersed entirely into aqueous phase. After centrifuging, the fine particles of extraction solvent are sedimented in the bottom of the conical test tube (5.0 +/- 0.2 microL). The performance of DLLME is illustrated with the determination of polycyclic aromatic hydrocarbons (PAHs) in water samples by using gas chromatography-flame ionization detection (GC-FID). Some important parameters, such as kind of extraction and disperser solvent and volume of them, and extraction time were investigated. Under the optimum conditions the enrichment factor ranged from 603 to 1113 and the recovery ranged from 60.3 to 111.3%. The linear range was 0.02-200 microg/L (four orders of magnitude) and limit of detection was 0.007-0.030 microg/L for most of analytes. The relative standard deviations (RSDs) for 2 microg/L of PAHs in water by using internal standard were in the range 1.4-10.2% (n = 5). The recoveries of PAHs from surface water at spiking level of 5.0 microg/L were 82.0-111.0%. The ability of DLLME technique in the extraction of other organic compounds such as organochlorine pesticides, organophosphorus pesticides and substituted benzene compounds (benzene, toluene, ethyl benzene, and xylenes) from water samples were studied. The advantages of DLLME method are simplicity of operation, rapidity, low cost, high recovery, and enrichment factor.
Sigma-Aldrich
Benzo[e]pyrene, 98%
Pricing and availability is not currently available.