Merck
  • Solvothermal synthesis of four polyoxometalate-based coordination polymers including diverse Ag(I)···π interactions.
Solvothermal synthesis of four polyoxometalate-based coordination polymers including diverse Ag(I)···π interactions.
Inorganic chemistry (2014-01-08)
Min-Xia Liang, Chan-Zi Ruan, Di Sun, Xiang-Jian Kong, Yan-Ping Ren, La-Sheng Long, Rong-Bin Huang, Lan-Sun Zheng
ABSTRACT
Four 3D POM-based silver coordination polymers, namely, [Ag17(ptz)11(PW12O40)2]n (1), [Ag17(ptz)11(PMo12O40)2]n (2), [Ag12(ptz)6(CN)2(SiW12O40)]n (3), and [Ag19(ptz)8(H2ptz)(H3ptz)(AgP5W30O110)·7H2O]n (4), have been obtained by solvothermal reaction of AgNO3 and 5-phenyl-1H-tetrazole (Hptz) ligand in the presence of four types of polyoxometalates. Structural analysis shows that four types of Ag(I)···π interactions, m-η(1), m/p-η(2), o/m-η(2), and o/m/p-η(3), were observed in compounds 1-4, depending on the polyoxometalates used. The in situ generated CN(-) ion in compound 3 shows unprecedented mixed σ and π bonding modes, similar to the C2(2-) ion in well-studied silver acetylides. For 4, the Na(+) ion in the Preyssler heteropolyoxoanion, [NaP5W30O110](14-), was exchanged by Ag(I) under solvothermal conditions, generating a novel [AgP5W30O110](14-) anion. In addition, the photoluminescence behavior of 1-4 was also investigated.
Sigma-Aldrich
5-Phenyl-1H-tetrazole, 99%