- Identification of CD137 as a potent monocyte survival factor.
Identification of CD137 as a potent monocyte survival factor.
Journal of leukocyte biology (1999-06-25)
J Langstein, H Schwarz
PMID10380906
ABSTRACT
CD137 (ILA/4-1BB), a member of the tumor necrosis factor (TNF) receptor family, promotes adherence and prolongs survival of human peripheral monocytes. It induces a strong expression of macrophage colony-stimulating factor (M-CSF), an essential monocyte survival factor. Monocyte survival induced by CD137 is primarily mediated by M-CSF and to a lesser extent by granulocyte-macrophage colony-stimulating factor and IL-3. Survival and induction of M-CSF are mediated via reverse signaling through a CD137 ligand expressed constitutively by peripheral monocytes.