Merck
  • The anaerobic decomposition of benzoic acid during methane fermentation. IV. Dearomatization of the ring and volatile fatty acids formed on ring rupture.
The anaerobic decomposition of benzoic acid during methane fermentation. IV. Dearomatization of the ring and volatile fatty acids formed on ring rupture.
Archives of microbiology (1978-08-01)
C L Keith, R L Bridges, L R Fina, K L Iverson, J A Cloran
PMID697507
ABSTRACT
A possible pathway for the anaerobic utilization of benzoic acid by a methanogenic consortium is suggested. Cyclohexane carboxylic acid and 1-cyclohexene-1-carboxylic acid have been identified as intermediates before ring rupture. Suprisingly, 3-cyclohexene-1-carboxylic acid interferes with utilization of other cyclic acids. In addition, isobutyric acid or short chain acids containing carbon-carbon double bonds could not be used without induction periods of a week or longer. A number of volatile fatty acids (heptanoic, valeric, butyric, propanoic, and acetic) have been identified and are suggested intermediates.
Sigma-Aldrich
1-Cyclohexene-1-carboxylic acid, 97%
Pricing and availability is not currently available.