Merck
  • Deoxygenation of some α-dicarbonyl compounds by tris(diethylamino)phosphine in the presence of fullerene C60.
Deoxygenation of some α-dicarbonyl compounds by tris(diethylamino)phosphine in the presence of fullerene C60.
The Journal of organic chemistry (2011-03-12)
Irina P Romanova, Andrey V Bogdanov, Vladimir F Mironov, Gulnara R Shaikhutdinova, Olga A Larionova, Shamil K Latypov, Alsu A Balandina, Dmitry G Yakhvarov, Aidar T Gubaidullin, Alina F Saifina, Oleg G Sinyashin
ABSTRACT
The reactions of such cyclic α-diketones as acenaphthenequinone, aceanthrenequinone, and N-alkylisatins, with hexaethyltriaminophosphine in the presence of the fullerene C(60), lead to the formation of methanofullerene derivatives under mild conditions. This process proceeds via deoxygenation of the dicarbonyl compound by the P(III) derivative and is likely to involve the intermediate formation of α-ketocarbenes. The structure of some methanofullerenes has been confirmed by NMR and XRD. The electrochemical behavior of the methanofullerenes was also investigated.
Sigma-Aldrich
Tris(dimethylamino)phosphine, 97%
Sigma-Aldrich
Aceanthrenequinone, 96%
Sigma-Aldrich
Tris(diethylamino)phosphine, 97%