Merck
  • Calcium-channel entry blocker therapy for hypertensive patients with concomitant renal impairment: a focus on isradipine.
Calcium-channel entry blocker therapy for hypertensive patients with concomitant renal impairment: a focus on isradipine.
Journal of clinical pharmacology (1994-12-01)
W H Frishman
ABSTRACT
In the treatment of hypertension in renally impaired patients, normalization of blood pressure alone may not be sufficient to prevent significant morbidity to the kidneys. Treatment must reduce pressure in the renal vasculature, otherwise glomerular filtration rate and renal plasma flow will continue to deteriorate. Isradipine a dihydropyridine calcium-channel blocker, has been investigated as a suitable treatment in this setting. Isradipine maintains glomerular filtration rate, preserves or enhances renal plasma flow, decreases renal vascular resistance, maintains or reduces filtration fraction, and exerts a sustained natriuretic effect, all of which may enable isradipine to slow the rate of progression of renal deterioration. In addition, isradipine may decrease proteinuria and may decrease glomerular capillary pressure by dilating both the efferent and afferent arterioles. Unlike older calcium-channel blockers, isradipine exhibits minimal cardiodepressant activity and is not associated with any negative inotropic effects. It is metabolized in the liver and dosage adjustments may not be necessary when administered to patients with renal insufficiency. Isradipine has a favorable renal effect profile and also has several properties that meet the requirements of other patient populations where an extra measure of antihypertensive safety is required, such as diabetics, dialysis patients, and transplant recipients. Side effects with isradipine are usually mild and transient, occurring in a dose-dependent manner.
Sigma-Aldrich
Isradipine, ≥98% (HPLC), solid