Merck
[Advance in latest studies on pharmacological effects of magnolol].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-03-13)
Yong Zhang, Fang Tang
ABSTRACT
Magnolol is one of main active constituents from Magnolia officinalis, a widely used traditional Chinese medicine. Previous studies have proved its multiple pharmacological effects, such as anti-oxidation, anti-microbial and anti-tumor. In recent years, more and more studies have focused on magnolol both at home and abroad. This essay summarizes the advance in the latest studies on the pharmacological effects of magnolol, and briefs main problems in current studies and future development orientation.
Sigma-Aldrich
Magnolol, ≥90% (HPLC), from plant
Magnolol, European Pharmacopoeia (EP) Reference Standard