Merck
Current methods for detecting ethylene in plants.
Annals of botany (2012-12-18)
Simona M Cristescu, Julien Mandon, Denis Arslanov, Jérôme De Pessemier, Christian Hermans, Frans J M Harren
ABSTRACT
In view of ethylene's critical developmental and physiological roles the gaseous hormone remains an active research topic for plant biologists. Progress has been made to understand the ethylene biosynthesis pathway and the mechanisms of perception and action. Still numerous questions need to be answered and findings to be validated. Monitoring gas production will very often complete the picture of any ethylene research topic. Therefore the search for suitable ethylene measuring methods for various plant samples either in the field, greenhouses, laboratories or storage facilities is strongly motivated. This review presents an update of the current methods for ethylene monitoring in plants. It focuses on the three most-used methods - gas chromatography detection, electrochemical sensing and optical detection - and compares them in terms of sensitivity, selectivity, time response and price. Guidelines are provided for proper selection and application of the described sensor methodologies and some specific applications are illustrated of laser-based detector for monitoring ethylene given off by Arabidopsis thaliana upon various nutritional treatments. Each method has its advantages and limitations. The choice for the suitable ethylene sensor needs careful consideration and is driven by the requirements for a specific application.
Sigma-Aldrich
Ethylene, puriss., ≥99.95% (GC)
Sigma-Aldrich
Ethylene, purum, ≥99.9%
Sigma-Aldrich
Ethylene, 99.99%
Sigma-Aldrich
Ethylene, ≥99.5%