Merck
  • Plasticity and conformational equilibria of influenza fusion peptides in model lipid bilayers.
Plasticity and conformational equilibria of influenza fusion peptides in model lipid bilayers.
Biochimica et biophysica acta (2014-01-21)
Neil R Haria, Luca Monticelli, Franca Fraternali, Christian D Lorenz
ABSTRACT
Membrane fusion is critical to eukaryotic cellular function and crucial to the entry of enveloped viruses such as influenza and human immunodeficiency virus. Influenza viral entry in the host cell is mediated by a 20-23 amino acid long sequence, called the fusion peptide. In the last years, possible structures for the fusion peptide and their implication in the membrane fusion initiation have been proposed; these ranging from an inverted V shaped α-helical structure to an α-helical hairpin, or to a complete α-helix. Here we develop a coarse grained approach to describe effectively the plasticity of the fusion peptide and the explored conformational states. We describe also a trimeric assembly for the fusion peptide and analyse the explored states in a 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine model membrane. For the single fusion peptide systems the kink angle observed experimentally for the V shaped structure shows a strong correlation with the orientation of the fusion peptide within the lipid bilayer. The trimeric fusion peptide model also experiences different conformational states and represents a more realistic model for the anchoring mechanism of one influenza haemagglutinin molecule. This article is part of a Special Issue entitled: Viral Membrane Proteins - Channels for Cellular Networking.
Sigma-Aldrich
2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, ≥99.0% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, ≥95.5% (GC), ≥98% (TLC)
Pricing and availability is not currently available.