Merck
  • Evaluation of secondary nitroalkanes, their nitronates, primary nitroalkanes, nitrocarbinols, and other aliphatic nitro compounds in the Ames Salmonella assay.
Evaluation of secondary nitroalkanes, their nitronates, primary nitroalkanes, nitrocarbinols, and other aliphatic nitro compounds in the Ames Salmonella assay.
Mutation research (1991-11-01)
C C Conaway, N S Hussain, B M Way, E S Fiala
ABSTRACT
The secondary nitroalkanes 2-nitropropane, 2-nitrobutane, 3-nitropentane and nitrocyclopentane, as well as their anionic forms (nitronates); the primary nitroalkanes 1-nitropropane, 1-nitrobutane, and 1-nitropentane and their respective nitronates; the nitrocarbinols 2-nitro-1-propanol, 2-nitro-1-butanol, 3-nitro-2-butanol, and 3-nitro-2-pentanol and their respective nitronates; 2-methyl-2-nitropropane, and 2-nitroso-2-nitropropane were tested in the Ames Salmonella assay using strains TA98, TA100 and TA102. Nitronates of the secondary nitroalkanes 2-nitropropane, 2-nitrobutane, 3-nitropentane, and nitrocyclopentane were significantly mutagenic in Salmonella strains TA100 and TA102 at 10-80 mumoles/plate, but the parent compounds were mutagenic at only a single dose level or were not mutagenic at all in the same dose range. The primary nitroalkanes and the nitrocarbinols were not mutagenic, or only marginally so, at the concentrations tested. The nitronates of the primary nitroalkanes and the nitrocarbinols reprotonated too rapidly under the conditions of the assay for adequate evaluation of mutagenicity. 2-Methyl-2-nitropropane was not mutagenic in strains TA100 and TA102; 2-nitroso-2-nitropropane was also not mutagenic in strains TA100 and TA102, but induced an equivocal mutagenic response in TA98. The positive Salmonella mutation data for the nitronates of the secondary nitroalkanes studied correlate very well with the very slow rate of reprotonation of secondary nitroalkane nitronates at pH 7.7 (Conaway et al. (1991) Cancer Res., 51, 3143), and provide further evidence that nitronates of secondary nitroalkanes, rather than the neutral parent forms with which they may be in equilibrium, are the more proximate mutagenic species.
Sigma-Aldrich
Nitrocyclopentane, 99%