Merck
  • Simultaneous detection of stable isotope-labeled and unlabeled L-tryptophan and of its main metabolites, L-kynurenine, serotonin and quinolinic acid, by gas chromatography/negative ion chemical ionization mass spectrometry.
Simultaneous detection of stable isotope-labeled and unlabeled L-tryptophan and of its main metabolites, L-kynurenine, serotonin and quinolinic acid, by gas chromatography/negative ion chemical ionization mass spectrometry.
Journal of mass spectrometry : JMS (2014-03-29)
Mitsue Sano, Véronique Ferchaud-Roucher, Charlotte Nael, Audrey Aguesse, Guillaume Poupeau, Blandine Castellano, Dominique Darmaun
ABSTRACT
A method for the detection of unlabeled and (15)N2 -labeled L-tryptophan (L-Trp), L-kynurenine (L-Kyn), serotonin (5-HT) and quinolinic acid (QA) in human and rat plasma by GC/MS is described. Labeled and unlabeled versions of these four products were analyzed as their acyl substitution derivatives using pentafluoropropionic anhydride and 2,2,3,3,3-pentafluoro-1-propanol. Products were then separated by GC and analyzed by selected ion monitoring using negative ion chemical ionization mass spectrometry. L-[(13)C11, (15)N2]-Trp, methyl-serotonin and 3,5-pyridinedicarboxylic acid were used as internal standards for this method. The coefficients of variation for inter-assay repeatability were found to be approximately 5.2% for L-Trp and (15)N2-Trp, 17.1% for L-Kyn, 16.9% for 5-HT and 5.8% for QA (n = 2). We used this method to determine isotope enrichments in plasma L-Trp over the course of a continuous, intravenous infusion of L-[(15) N2 ]Trp in pregnant rat in the fasting state. Plasma (15)N2-Trp enrichment reached a plateau at 120 min. The free Trp appearance rate (Ra) into plasma was 49.5 ± 3.35 µmol/kg/h. The GC/MS method was applied to determine the enrichment of (15)N-labeled L-Trp, L-Kyn, 5-HT and QA concurrently with the concentration of non-labeled L-Trp, L-Kyn, 5-HT and QA in plasma. This method may help improve our understanding on L-Trp metabolism in vivo in animals and humans and potentially reveal the relative contribution of the four pathways of L-Trp metabolism.
Sigma-Aldrich
2,2,3,3,3-Pentafluoro-1-propanol, 97%
Pricing and availability is not currently available.