Merck
  • Antioxidant and hepatoprotective activity of punicalagin and punicalin on carbon tetrachloride-induced liver damage in rats.
Antioxidant and hepatoprotective activity of punicalagin and punicalin on carbon tetrachloride-induced liver damage in rats.
The Journal of pharmacy and pharmacology (1998-08-28)
C C Lin, Y F Hsu, T C Lin, F L Hsu, H Y Hsu
ABSTRACT
Punicalagin and punicalin, isolated from the leaves of Terminalia catappa L., are used to treat dermatitis and hepatitis. Both compounds have strong antioxidative activity. The antihepatotoxic activity of punicalagin and punicalin on carbon tetrachloride (CCl4)-induced toxicity in the rat liver was evaluated. Levels of serum glutamate-oxalate-transaminase and glutamate-pyruvate-trans-aminase were increased by administration of CCl4 and reduced by drug treatment. Histological changes around the liver central vein and oxidation damage induced by CCl4 also benefited from drug treatment. The results show that both punicalagin and punicalin have anti-hepatotoxic activity but that the larger dose of punicalin induced liver damage. Thus even if tannins have strong antioxidant activity at very small doses, treatment with a larger dose will induce cell damage.
Product Description
Supelco
Punicalin, mixture of anomers, analytical standard