Merck
  • Trimethoprim-sulfamethoxazole treatment does not reverse obstructive pulmonary changes in pneumocystis-colonized nonhuman primates with SHIV infection.
Trimethoprim-sulfamethoxazole treatment does not reverse obstructive pulmonary changes in pneumocystis-colonized nonhuman primates with SHIV infection.
Journal of acquired immune deficiency syndromes (1999) (2013-10-15)
Heather M Kling, Timothy W Shipley, Siobhan Guyach, Rebecca Tarantelli, Alison Morris, Karen A Norris
ABSTRACT
Despite antiretroviral therapy and trimethoprim-sulfamethoxazole (TMP-SMX) prophylaxis, Pneumocystis pneumonia remains an important serious opportunistic infection in HIV-infected persons. Pneumocystis (Pc) colonization in HIV-infected individuals and in HIV-uninfected smokers is associated with chronic obstructive pulmonary disease (COPD). We previously developed a nonhuman primate model of HIV infection and Pc colonization and demonstrated that Pc colonization correlated with COPD development. In the present study, we examined kinetics of COPD development in non-human primate and tested the effect of Pc burden reduction on pulmonary function by TMP-SMX treatment. Cynomolgus macaques (n = 16) were infected with simian/human immunodeficiency virus (SHIV89.6P), and natural Pc colonization was examined by nested polymerase chain reaction of serial bronchoalveolar lavage fluid and anti-Pc serology. Eleven of 16 monkeys became Pc colonized by 16 weeks post simian-human immunodeficiency virus (SHIV) infection. Pc colonization of SHIV-infected monkeys led to progressive declines in pulmonary function as early as 4 weeks after Pc detection. SHIV-infected and Pc-negative monkeys maintained normal lung function. At 25 weeks post-SHIV infection, TMP-SMX treatment was initiated in 7 Pc-positive (Pc+) (TMP: 20 mg/kg and SMX: 100 mg/kg, daily for 48 weeks) and 5 Pc-negative (Pc-) monkeys. Four SHIV+/Pc+ remained untreated for the duration of the experiment. Detection frequency of Pc in serial bronchoalveolar lavage fluid (P < 0.001), as well as plasma Pc antibody titers (P = 0.02) were significantly reduced in TMP-SMX-treated macaques compared with untreated. Reduction of Pc colonization by TMP-SMX treatment did not improve pulmonary function, supporting the concept that Pc colonization results in early, permanent obstructive changes in the lungs of immunosuppressed macaques.