Merck
  • Recent advances on physiological functions and biotechnological production of epilactose.
Recent advances on physiological functions and biotechnological production of epilactose.
Applied microbiology and biotechnology (2013-01-16)
Wanmeng Mu, Qiuxi Li, Chen Fan, Chen Zhou, Bo Jiang
ABSTRACT
Epilactose (4-O-β-D-galactopyranosyl-D-mannose), an epimer of lactose, is a rare disaccharide existing extremely small quantities in heat-treated milk, in which epilactose is produced by non-enzymatic catalysis from lactose. This disaccharide is a kind of non-digestible carbohydrate, has a good prebiotic effect, and promotes intestinal mineral absorption. This article presents a review of recent studies on epilactose formation in food system, qualitative and quantitative analysis, and its physiological functions. In addition, the biochemical properties and kinetic parameters of the epilactose-producing enzyme, cellobiose 2-epimerase, are compared, and the biotechnological production of epilactose from lactose is reviewed.
Product Description
USP
Epilactose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
4-O-β-Galactopyranosyl-D-mannopyranose, ≥98.0% (HPLC)