Merck
  • Effect of sedation with detomidine on intraocular pressure with and without topical anesthesia in clinically normal horses.
Effect of sedation with detomidine on intraocular pressure with and without topical anesthesia in clinically normal horses.
Journal of the American Veterinary Medical Association (2012-01-20)
Dana L Holve
ABSTRACT
To determine the effect of sedation with detomidine on intraocular pressure (IOP) in standing horses and whether topical ocular application of anesthetic alters this effect. Clinical trial. 15 clinically normal horses. Horses were assigned to group 1 (n = 7) or 2 (8). Intraocular pressure measurements were obtained at baseline (before sedation) and 10 minutes after IV administration of detomidine (0.02 mg/kg [0.009 mg/lb]). Group 1 horses had an additional IOP measurement at 20 minutes after sedation. For group 2 horses, topical ocular anesthetic was administered 10 minutes prior to baseline IOP measurements. Mean ± SD baseline IOP for horses without topical anesthesia (group 1 horses; IOP, 24.30 ± 3.09 mm Hg) was significantly lower, compared with that of horses with topical anesthesia (group 2 horses; IOP, 30.40 ± 3.25 mm Hg). Compared with baseline values, IOP at 10 minutes after sedation decreased significantly in all horses (by 3.61 ± 1.48 mm Hg and 5.78 ± 4.32 mm Hg in groups 1 and 2, respectively). In group 1 horses, IOP at 10 (20.69 ± 3.45 mm Hg) and 20 (19.96 ± 2.13 mm Hg) minutes after sedation was significantly decreased, compared with baseline values; however, the difference between IOP at 10 versus 20 minutes was not significant. The difference in IOP between group 1 and 2 horses at 10 minutes after sedation was not significant. IV administration of detomidine caused a decrease in IOP in clinically normal horses and may be a safe sedative when performing ocular procedures.
Product Description
Detomidine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.