Merck
  • [Effects of carteolol hydrochloride on experimental choroidal neovascularization].
[Effects of carteolol hydrochloride on experimental choroidal neovascularization].
Nippon Ganka Gakkai zasshi (2011-05-24)
Susumu Ishida, Hiroshi Mochimaru, Norihiro Nagai, Kousuke Noda, Yoko Ozawa
ABSTRACT
To investigate the effects of carteolol hydrochloride on choroidal neovascularization (CNV). Laser photocoagulation was performed to induce CNV in C 57 BL/6 mice. The response of CNV was assessed by fluorescein isothiocyanate-isolectin B 4 staining. The expression of intercellular adhesion molecule (ICAM)-1 and monocyte chemotactic protein (MCP)-1 in the retinal pigment epithelium (RPE)-choroid complex and tumor necrosis factor (TNF)-alpha in the RAW 264.7 macrophage culture was evaluated by enzyme-linked immunosorbent assay. Carteolol hydrochloride application led to significant suppression of the generation of CNV, the production of ICAM-1 and MCP-1 in the RPE-choroid, and macrophage expression of TNF-alpha. Carteolol hydrochloride prevented CNV development through its anti-inflammatory action.