Merck
  • [Chemical constituents of antibacterial activity fraction of Angelica polymorpha].
[Chemical constituents of antibacterial activity fraction of Angelica polymorpha].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2008-12-31)
Zheng-jun Cai, Fei-jun Dan, Fan Cheng, Jun-zhi Wang, Kun Zou
ABSTRACT
To study chemical constituents of antibacterial activity fraction of Angelica polymorpha. Compounds were isolated by repeatedly silica gel column chromatography and recrystallization. Their structures were identified by physical and chemical evidences and spectral methods. Seven compounds were obtained from the antibacterial activity fraction, their structures were elucidated as: bisabolangelone(I), isoimperatorin (II), oxypeucedanine(III), isooxypeucedanine(IV), oxypeucedanin hydrate(V), bergapten(VI), pabulenol(VII). Bisabolangelone(I) is obtained from this plant for the first time. Compound (II)-(VII) belong to linear furanocourmarins.
Product Description
Supelco
Isoimperatorin, analytical standard