Merck
  • A possible role for cysteinyl-leukotrienes in non-ionic contrast media induced adverse reactions.
A possible role for cysteinyl-leukotrienes in non-ionic contrast media induced adverse reactions.
European journal of radiology (2005-09-01)
Ingrid Böhm, Ulrich Speck, Hans Schild
ABSTRACT
To test whether mono- or dimeric X-ray contrast media (CM) may induce the de novo production of cysteinyl-leukotriens (cys-LT), that could contribute to allergic/allergy-like side effects. Leukocytes from 39 patients receiving iopromide or iotrolan for routine CT-examination were analyzed for the production of cys-LT. Histamine levels were analyzed in plasma specimens. One patient with a positive history of a previous CM-reaction did not receive CM-injection. Three patients of the iopromide and five of the iotrolan group showed adverse reactions. Reactors had increased cys-LT values in samples obtained before CM-injection induced by the positive control (anti-FcepsilonRI antibodies) (6763.7 pg/ml+/-1367.3 versus 2299.8 pg/ml+/-399.2; p<0.007). Patients with versus without CM-reaction did not differ significantly with respect to their histamine values before CM-administration. In vitro iopromide (p<0.0002) and iotrolan (p<0.0008) induced significant cys-LT production as compared to IL-3 stimulation. In vivo both CM induced a significant increase 6h after CM administration (p<0.05). Our findings suggest that both CM seem to induce cys-LT production. As to whether the observed increased values in pre-dose samples of patients with as compared to those without reactions could contribute to identify high risk patients should be investigated in larger patient groups in future.
Product Description
Iotrolan, European Pharmacopoeia (EP) Reference Standard