- Dipropyl 3,6-diphenyl-1,2-dihydro-1,2,4,5-tetrazine-1,2-dicarboxylate.
Dipropyl 3,6-diphenyl-1,2-dihydro-1,2,4,5-tetrazine-1,2-dicarboxylate.
Acta crystallographica. Section C, Crystal structure communications (2003-05-14)
Guo-Wu Rao, Wei-Xiao Hu
PMID12743415
ABSTRACT
The title compound, C(22)H(24)N(4)O(4), was prepared from propyl chloroformate and 3,6-diphenyl-1,2-dihydro-s-tetrazine. This reaction yields the title compound rather than dipropyl 3,6-diphenyl-1,4-dihydro-s-tetrazine-1,4-dicarboxylate. The 2,3-diazabutadiene group in the central six-membered ring is not planar; the C=N double-bond length is 1.285 (2) A, and the average N-N single-bond length is 1.401 (3) A, indicating a lack of conjugation. The ring has a twist conformation, in which adjacent N atoms lie +/- 0.3268 (17) A from the plane of the ring. The molecule has twofold crystallographic symmetry.
MATERIALS