Merck
  • Measurement of aromatisation by a urine technique suitable for the evaluation of aromatase inhibitors in vivo.
Measurement of aromatisation by a urine technique suitable for the evaluation of aromatase inhibitors in vivo.
Journal of enzyme inhibition (1991-01-01)
S Jacobs, P E Lønning, B Haynes, L Griggs, M Dowsett
ABSTRACT
By modification of a recently developed method for separation of radio-labelled urinary oestrogens we were able to separate oestrogen metabolites and measure their isotope ratios in urine following injections of [3H]delta 4-androstenedione and [14C]oestrone. This method provides a useful tool for studying in vivo aromatisation of delta 4-androstenedione into oestrone in breast cancer patients before and during treatment with aromatase inhibitors.
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3, aqueous solution, ≥90,000 units/mL
Pricing and availability is not currently available.