Merck
  • Rhomboid protease PARL mediates the mitochondrial membrane potential loss-induced cleavage of PGAM5.
Rhomboid protease PARL mediates the mitochondrial membrane potential loss-induced cleavage of PGAM5.
The Journal of biological chemistry (2012-08-24)
Shiori Sekine, Yusuke Kanamaru, Masato Koike, Ayako Nishihara, Masahiro Okada, Hideyuki Kinoshita, Miki Kamiyama, Junichi Maruyama, Yasuo Uchiyama, Naotada Ishihara, Kohsuke Takeda, Hidenori Ichijo
ABSTRACT
Regulated intramembrane proteolysis is a widely conserved mechanism for controlling diverse biological processes. Considering that proteolysis is irreversible, it must be precisely regulated in a context-dependent manner. Here, we show that phosphoglycerate mutase 5 (PGAM5), a mitochondrial Ser/Thr protein phosphatase, is cleaved in its N-terminal transmembrane domain in response to mitochondrial membrane potential (ΔΨ(m)) loss. This ΔΨ(m) loss-dependent cleavage of PGAM5 was mediated by presenilin-associated rhomboid-like (PARL). PARL is a mitochondrial resident rhomboid serine protease and has recently been reported to mediate the cleavage of PINK1, a mitochondrial Ser/Thr protein kinase, in healthy mitochondria with intact ΔΨ(m). Intriguingly, we found that PARL dissociated from PINK1 and reciprocally associated with PGAM5 in response to ΔΨ(m) loss. These results suggest that PARL mediates differential cleavage of PINK1 and PGAM5 depending on the health status of mitochondria. Our data provide a prototypical example of stress-dependent regulation of PARL-mediated regulated intramembrane proteolysis.