Merck
  • Preparation of DL-2,3,4-trihydroxybutylarsonic acid and dl-2,3-dihydroxybutane-1,4-bis(arsonic acid): starting compounds for novel arsonolipids.
Preparation of DL-2,3,4-trihydroxybutylarsonic acid and dl-2,3-dihydroxybutane-1,4-bis(arsonic acid): starting compounds for novel arsonolipids.
Chemistry and physics of lipids (2007-06-09)
Gerasimos M Tsivgoulis, Maria A Lala, Panayiotis V Ioannou
ABSTRACT
The reaction of DL-1,3-butadiene diepoxide and of DL-1,4-dibromo-2,3-butanediol with aqueous alkaline sodium arsenite, "Na(3)AsO(3)", gave mixtures of the title arsonic acids which can be separated by anion exchange resin. Characterization of by-products leads to a better understanding of these reactions. These compounds are valuable intermediates for the preparation of novel arsonic acids and bis(arsonic acids).
Sigma-Aldrich
1,4-Dibromo-2,3-butanediol, 95%, mixture of R,R and S,S isomers