Merck
  • Arabidopsis DNA methyltransferase AtDNMT2 associates with histone deacetylase AtHD2s activity.
Arabidopsis DNA methyltransferase AtDNMT2 associates with histone deacetylase AtHD2s activity.
Biochemical and biophysical research communications (2010-03-25)
Yuan Song, Keqiang Wu, Sangeeta Dhaubhadel, Lizhe An, Lining Tian
ABSTRACT
DNA methyltransferase2 (DNMT2) is always deemed to be enigmatic, because it contains highly conserved DNA methyltransferase motifs but lacks the DNA methylation catalytic capability. Here we show that Arabidopsis DNA methyltransferase2 (AtDNMT2) is localized in nucleus and associates with histone deacetylation. Bimolecular fluorescence complementation and pull-down assays show AtDNMT2 interacts with type-2 histone deacetylases (AtHD2s), a unique type of histone deacetylase family in plants. Through analyzing the expression of AtDNMT2: ss-glucuronidase (GUS) fusion protein, we demonstrate that AtDNMT2 has the ability to repress gene expression at transcription level. Meanwhile, the expression of AtDNMT2 gene is altered in athd2c mutant plants. We propose that AtDNMT2 possibly involves in the activity of histone deacetylation and plant epigenetic regulatory network.
Sigma-Aldrich
Plant/Fungi RNA Purification Kit, sufficient for 50 purifications