Merck
  • Regio- and diastereoselective synthesis of 2-alkylidenetetrahydrofurans by domino SN/SN' and SN/SN reactions of 1,3-dicarbonyl dianions.
Regio- and diastereoselective synthesis of 2-alkylidenetetrahydrofurans by domino SN/SN' and SN/SN reactions of 1,3-dicarbonyl dianions.
The Journal of organic chemistry (2001-09-01)
P Langer, E Holtz, I Karimé, N N Saleh
ABSTRACT
The domino C,O-cyclodialkylation reaction of dilithiated 1,3-dicarbonyl compounds with 1,4-dibromo-2-butene resulted in regio- and diastereoselective formation of 2-alkylidene-5-vinyltetrahydrofurans. The cyclization of 1,3-dicarbonyl dianions with 1-bromo-2-chloroethane regio- and diastereoselectively afforded 2-alkylidenetetrahydrofurans under thermodynamic reaction control.
Sigma-Aldrich
1-Bromo-2-chloroethane, 98%
Pricing and availability is not currently available.