Merck
  • Synthesis of high silicon content SAPO(4)-5 using anionic surfactants in a hexanol/aqueous two phase media.
Synthesis of high silicon content SAPO(4)-5 using anionic surfactants in a hexanol/aqueous two phase media.
Chemical communications (Cambridge, England) (2002-07-18)
Xue S Han, Craig D Williams, Darren F Lee, Catherine I Round
ABSTRACT
High silicon content SAPO(4)-5 (up to 0.511 atoms per unit cell) has been synthesised, using sodium 3-bromopropanesulfonate, sodium 1-butanesulfonate, sodium naphthalene-1-sulfonate or sodium n-decyl sulfate as surfactants; the SiO2 in the reaction gels ranged up to 3.0 (molar ratio), silicon incorporation was confirmed by XRD, XRF, TG-DTA, FT-IR and SEM techniques.
Sigma-Aldrich
Sodium 1-butanesulfonate, 98%
Pricing and availability is not currently available.