- Design, synthesis and structure-activity relationship of novel RXR-selective modulators.
Design, synthesis and structure-activity relationship of novel RXR-selective modulators.
Bioorganic & medicinal chemistry letters (2004-03-10)
Pierre-Yves Michellys, Jennifer D'Arrigo, Timothy A Grese, Donald S Karanewsky, Mark D Leibowitz, Dale A Mais, Christopher M Mapes, Anne Reifel-Miller, Deepa Rungta, Marcus F Boehm
PMID15006411
ABSTRACT
The synthesis and in vitro characterization of novel RXR-selective ligands possessing various substituted 1-benzofuran or 1-benzothiophene moieties are described.