Merck
  • Can long-term follow-up strategies be determined using a nomogram-based prediction model of malignancy among intraductal papillary mucinous neoplasms of the pancreas?
Can long-term follow-up strategies be determined using a nomogram-based prediction model of malignancy among intraductal papillary mucinous neoplasms of the pancreas?
Pancreas (2014-03-14)
Susumu Hijioka, Yasuhiro Shimizu, Nobumasa Mizuno, Kazuo Hara, Hiroshi Imaoka, Mohamed A Mekky, Vikram Bhatia, Yoshikuni Nagashio, Toshiyuki Hasegawa, Akihide Shinagawa, Masanari Sekine, Masahiro Tajika, Tsutomu Tanaka, Makoto Ishihara, Yasumasa Niwa, Kenji Yamao
ABSTRACT
This study investigated whether a risk assessment nomogram can predict the malignant potential of intraductal papillary mucinous neoplasms (IPMNs) and provide valuable information for the follow-up and counseling strategies of such patients. We studied 126 of 589 patients with IPMN who were followed up for at least 36 months with annual endoscopic ultrasonography. We analyzed scores derived from our nomogram, incorporating the parameters of sex, lesion type, mural nodule height, and pancreatic juice cytology determined at the initial IPMN evaluation. The rate of malignant IPMNs was 5.5% (7/126). The initial average nomogram score was 19.8 (range, 0-55), and the final follow-up average was 23.8 (range, 0-109). When a cutoff score was set at 35 points, the sensitivity, specificity, and accuracy of the nomogram to assess malignancy risk were 87.5%, 96.6%, and 96%, respectively. The area under the receiver operating characteristic curve of malignant IPMN prediction during follow-up was 0.865. The ability of the nomogram to predict malignancy in patients with IPMN was validated. Our findings can suggest that a follow-up for patients at high and low risk for cancer progression could be scheduled every 3 to 6 and 12 months, respectively.
Sigma-Aldrich
Chalcone 4 hydrate, ≥98% (HPLC)