Merck
Dual-modality gene reporter for in vivo imaging.
Proceedings of the National Academy of Sciences of the United States of America (2013-12-19)
P Stephen Patrick, Jayne Hammersley, Louiza Loizou, Mikko I Kettunen, Tiago B Rodrigues, De-En Hu, Sui-Seng Tee, Robin Hesketh, Scott K Lyons, Dmitry Soloviev, David Y Lewis, Silvio Aime, Sandra M Fulton, Kevin M Brindle
ABSTRACT
The ability to track cells and their patterns of gene expression in living organisms can increase our understanding of tissue development and disease. Gene reporters for bioluminescence, fluorescence, radionuclide, and magnetic resonance imaging (MRI) have been described but these suffer variously from limited depth penetration, spatial resolution, and sensitivity. We describe here a gene reporter, based on the organic anion transporting protein Oatp1a1, which mediates uptake of a clinically approved, Gd(3+)-based, hepatotrophic contrast agent (gadolinium-ethoxybenzyl-diethylenetriamine pentaacetic acid). Cells expressing the reporter showed readily reversible, intense, and positive contrast (up to 7.8-fold signal enhancement) in T1-weighted magnetic resonance images acquired in vivo. The maximum signal enhancement obtained so far is more than double that produced by MRI gene reporters described previously. Exchanging the Gd(3+) ion for the radionuclide, (111)In, also allowed detection by single-photon emission computed tomography, thus combining the spatial resolution of MRI with the sensitivity of radionuclide imaging.
Sigma-Aldrich
Gadolinium, foil, thickness 1 mm, 99.9% trace rare earth metals basis
Sigma-Aldrich
Gadolinium, chips
Sigma-Aldrich
Gadolinium, −40 mesh, 99% trace rare earth metals basis
Sigma-Aldrich
Gadolinium, ingot, 99.9% trace rare earth metals basis