Merck
  • Cu-NHC-TEMPO catalyzed aerobic oxidation of primary alcohols to aldehydes.
Cu-NHC-TEMPO catalyzed aerobic oxidation of primary alcohols to aldehydes.
The Journal of organic chemistry (2013-08-16)
Xiaolong Liu, Qinqin Xia, Yuejiao Zhang, Congyan Chen, Wanzhi Chen
ABSTRACT
Imidazolium salts bearing TEMPO groups react with commercially available copper powder affording Cu-NHC complexes. The in situ generated Cu-NHC-TEMPO complexes are quite efficient catalysts for aerobic oxidation of primary alcohols into aldehydes. The catalyst is easily available, and various primary alcohols were selectively converted to aldehydes in excellent yields.
Sigma-Aldrich
TEMPO, 98%
Sigma-Aldrich
TEMPO, purified by sublimation, 99%