Merck
  • Batch investigations on P immobilization from wastewaters and sediment using natural calcium rich sepiolite as a reactive material.
Batch investigations on P immobilization from wastewaters and sediment using natural calcium rich sepiolite as a reactive material.
Water research (2013-06-15)
Hongbin Yin, Ming Kong, Chengxin Fan
ABSTRACT
Phosphorus from wastewaters and sediment flux to surface water represents a major source of lake eutrophication. Active filtration and in situ capping (which refers to placement of a covering or cap over an in-situ deposit of contaminated sediment) are widely used as a means to immobilize phosphorus from wastewaters and sediment, to mitigate lake eutrophication. There is, however, a need to develop more efficient means of immobilizing phosphorus through the development of binding agents. In this study, natural calcium-rich sepiolite (NCSP) was calcined at a range of temperatures, to enhance its phosphorus removal capacity. Batch studies showed that the 900 °C calcinated NCSP (NCSP900) exhibited excellent sorption performance, attaining a phosphorus removal efficiency of 80.0%-99.9% in the range of 0.05 mg/L-800 mg/L phosphorus concentrations with a dosage of 20 g/L. The material displayed rapid sorption rate (maximum amount of 99.9% of phosphate removal with 5 min) and could lower the very high phosphate concentration (200 mg/L) to less than 0.1 mg/L after 4 h adsorption. It was also noted that factors such as pH, competing anions (except [Formula: see text] ) and humic acid, had no effect on phosphorus removal capacity. The sediment immobilization experiment indicated that NCSP900 had the capacity to transform reactive phosphorus into inert-phosphorus and significantly reduce the amount of algal-bioavailable phosphorus. The excellent phosphorus binding performance of NCSP900 was mainly due to the improvement of point of zero charge (pHPZC) as well as the transformation of the inert-calcium of NCSP to active free CaO during calcination. Phosphorus speciation indicated that phosphorus was mainly captured by relatively stable calcium-bound phosphorus (Ca-P) precipitation, which can account for 80.1% of the total phosphorus. This study showed that NCSP900 could be used as an efficient binding agent for the sequestration of phosphorus from wastewaters and sediment.
Sigma-Aldrich
Magnesium trisilicate hydrate, tested according to Ph. Eur.