Merck
  • Cross-coupling reactions of alkenylsilanolates. Investigation of the mechanism and identification of key intermediates through kinetic analysis.
Cross-coupling reactions of alkenylsilanolates. Investigation of the mechanism and identification of key intermediates through kinetic analysis.
Journal of the American Chemical Society (2004-04-15)
Scott E Denmark, Ramzi F Sweis
ABSTRACT
The mechanism of the fluoride-free, palladium-catalyzed cross-coupling reaction of potassium (E)-heptenyldimethylsilanolate, K(+)(E)-1(-), with 2-iodothiophene has been investigated through kinetic analysis. The order of each component was determined by plotting the initial rates of the reaction against concentration. These data provided a mechanistic picture which involves a fast and irreversible oxidative insertion of palladium into the aryl iodide and a subsequent intramolecular transmetalation step from a complex containing a silicon-oxygen-palladium linkage. First-order behavior at low concentrations of silanolate with excess palladium(0) complex supports the formation of this complex as the turnover-limiting step. The change to zeroth-order dependence on silanolate at high concentrations is consistent with the intramolecular transmetalation becoming the turnover-limiting step.
Sigma-Aldrich
2-Iodothiophene, 98%