Merck
  • Neutral and acidic products derived from hydroxyl radical-induced oxidation of arabinotriose assessed by electrospray ionisation mass spectrometry.
Neutral and acidic products derived from hydroxyl radical-induced oxidation of arabinotriose assessed by electrospray ionisation mass spectrometry.
Journal of mass spectrometry : JMS (2014-04-11)
Ana S P Moreira, Elisabete V da Costa, Dmitry V Evtuguin, Manuel A Coimbra, Fernando M Nunes, M Rosário M Domingues
ABSTRACT
The oxidation of α-(1 → 5)-L-arabinotriose (Ara3), an oligosaccharide structurally related to side chains of coffee arabinogalactans, was studied in reaction with hydroxyl radicals generated under conditions of Fenton reaction (Fe(2+)/H2O2). The acidic and neutral oxidation products were separated by ligand exchange/size-exclusion chromatography, subsequently identified by electrospray ionisation mass spectrometry (ESI-MS) and structurally characterised by tandem MS (ESI-MS/MS). In acidic fraction were identified several oxidation products containing an acidic residue at the corresponding reducing end of Ara3, namely arabinonic acid, and erythronic, glyceric and glycolic acids formed by oxidative scission of the furanose ring. In neutral fractions were identified derivatives containing keto, hydroxy and hydroperoxy moieties, as well as derivatives resulting from the ring scission at the reducing end of Ara3. In both acidic and neutral fractions, beyond the trisaccharide derivatives, the corresponding di- and monosaccharide derivatives were identified indicating the occurrence of oxidative depolymerisation. The structural characterisation of these oxidation products by ESI-MS/MS allowed the differentiation of isobaric and isomeric species of acidic and neutral character. The species identified in this study may help in detection of roasting products associated with the free radical-mediated oxidation of coffee arabinogalactans.
Sigma-Aldrich
L-Arabino-1,4-lactone, ≥95.0% (GC)