- Alcohol cross-coupling reactions catalyzed by Ru and Ir terpyridine complexes.
Alcohol cross-coupling reactions catalyzed by Ru and Ir terpyridine complexes.
Organic & biomolecular chemistry (2008-11-14)
Dinakar Gnanamgari, Chin Hin Leung, Nathan D Schley, Sheena T Hilton, Robert H Crabtree
PMID19005605
ABSTRACT
Primary alcohols can be coupled with secondary benzylic alcohols by an air-stable catalytic system involving terpyridine ruthenium or iridium complexes.
MATERIALS