Merck
  • Mechanistic insights into oxidation of 2-methyl-1-naphthol with dioxygen: autoxidation or a spin-forbidden reaction?
Mechanistic insights into oxidation of 2-methyl-1-naphthol with dioxygen: autoxidation or a spin-forbidden reaction?
The journal of physical chemistry. B (2011-09-15)
Oxana A Kholdeeva, Irina D Ivanchikova, Olga V Zalomaeva, Alexander B Sorokin, Igor Y Skobelev, Eugenii P Talsi
ABSTRACT
Oxidation of 2-methyl-1-naphthol (MNL) with molecular oxygen proceeds efficiently under mild reaction conditions (3 atm O(2), 60-80 °C) in the absence of any catalyst or sensitizer and produces 2-methyl-1,4-naphthoquinone (MNQ, menadione, or vitamin K(3)) with selectivity up to 80% in nonpolar solvents. (1)H NMR and (1)H,(1)H-COSY studies revealed the formation of 2-methyl-4-hydroperoxynaphthalene-1(4H)-one (HP) during the reaction course. Several mechanistic hypotheses, including conventional radical autoxidation, electron transfer mechanisms, photooxygenation, and thermal intersystem crossing (ISC), have been evaluated using spectroscopic, mass-spectrometric, spin-trapping, (18)O(2) labeling, kinetic, and computational techniques. Several facts collectively implicate that ISC contributes significantly into MNL oxidation with O(2) at elevated pressure: (i) the reaction rate is unaffected by light; (ii) C-C-coupling dimers are practically absent; (iii) the reaction is first order in both MNL and O(2); (iv) the observed activation parameters (ΔH(‡) = 8.1 kcal mol(-1) and ΔS(‡) = -50 eu) are similar to those found for the spin-forbidden oxidation of helianthrene with (3)O(2) (Seip, M.; Brauer, H.-D. J. Am. Chem. Soc.1992, 114, 4486); and (v) the external heavy atom effect (2-fold increase of the reaction rate in iodobenzene) points to spin inversion in the rate-limiting step.
Sigma-Aldrich
2-Methyl-1-naphthol, 98%