Merck
  • Conformationally restricted indolopiperidine derivatives as potent CCR2B receptor antagonists.
Conformationally restricted indolopiperidine derivatives as potent CCR2B receptor antagonists.
Bioorganic & medicinal chemistry letters (2001-08-22)
J Witherington, V Bordas, D G Cooper, I T Forbes, A D Gribble, R J Ife, T Berkhout, J Gohil, P H Groot
ABSTRACT
The preparation and biological evaluation of a series of indolopiperidine CCR2B receptor antagonists possessing a conformationally restricted C-5 linker chain in combination with a restricted piperidine ring are described. Compared to the parent compound 1, analogue 8 shows a dramatic improvement in selectivity against a range of 5-HT and dopaminergic receptors.
Sigma-Aldrich
3,4-Dichlorocinnamic acid, 97%