Merck
Dimethylthiocarbamate (DMTC): an alcohol protecting group.
Organic letters (2003-12-05)
D K Barma, A Bandyopadhyay, Jorge H Capdevila, J R Falck
ABSTRACT
Dimethylthiocarbamates (DMTCs), prepared from the corresponding alcohols using commercial dimethylthiocarbamoyl chloride, are spectrally simple, achiral, and nonpolar. DMTCs are moderately to highly stable to a wide range of reagents and conditions including metal hydrides, hydroboration, ylides, NaOH, HCl, organolithiums, Grignards, DDQ, PCC, Swern, n-Bu(4)NF, CrCl(2), heat, and Lewis acids. They are readily removed by NaIO(4) or H(2)O(2) in the presence of other common alcohol protecting groups. [structure: see text]
Sigma-Aldrich
Dimethylthiocarbamoyl chloride, 97%