Merck
Tuning the size of macrocyclic cavities in trianglimine macrocycles.
Organic & biomolecular chemistry (2005-05-13)
Nikolai Kuhnert, Nicolai Burzlaff, Chirag Patel, Ana Lopez-Periago
ABSTRACT
The synthesis of aromatic dicarboxaldehydes is described along with their reactivity in the [3 + 3] cyclocondensation reaction with (1R,2R)-diaminocyclohexane to give trianglimine macrocycles. In particular, the scope and limitation of the reaction with regard to complete control of the cavity size of the macrocycles is discussed producing a total of 11 macrocycles with different cavity sizes ranging from 9 to 23 angstroms.
Sigma-Aldrich
1,4-Dibromo-2,5-dimethylbenzene, 98%
Pricing and availability is not currently available.