Merck
  • Sensitive determination of pethidine in body fluids by gas chromatography-tandem mass spectrometry.
Sensitive determination of pethidine in body fluids by gas chromatography-tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2003-06-28)
Akira Ishii, Miwa Tanaka, Rina Kurihara, Kanako Watanabe-Suzuki, Takeshi Kumazawa, Hiroshi Seno, Osamu Suzuki, Yoshinao Katsumata
ABSTRACT
We have presented a simple and sensitive method for determining pethidine, a narcotic analgesic drug in body fluids by gas chromatography-tandem mass spectrometry (GC-MS/MS). Pethidine and 4'-piperidinoacetophenone (internal standard) were extracted from body fluids with Bond Elut C(18) columns; the recoveries were above 85% for both compounds. The calibration curves for blood and urine showed good linearities in the range of 1.25-40 ng/ml. Its detection limits (signal-to-noise ratio=3) were estimated to be approximately 0.5 ng/ml of whole blood and urine.
Sigma-Aldrich
4′-Piperidinoacetophenone, 97%