- Human Smad3 and Smad4 are sequence-specific transcription activators.
Human Smad3 and Smad4 are sequence-specific transcription activators.
Molecular cell (1998-07-14)
L Zawel, J L Dai, P Buckhaults, S Zhou, K W Kinzler, B Vogelstein, S E Kern
PMID9660945
ABSTRACT
Mounting evidence indicates that Smad proteins are required for TGF beta signaling, but the way(s) in which Smad proteins propagate this signal is unclear. We found that two human Smad proteins (Smad3 and Smad4) could specifically recognize an identical 8 bp palindromic sequences (GTCTAGAC). Tandem repeats of this palindrome conferred striking TGF beta responsiveness to a minimal promoter. This responsiveness was abrogated by targeted deletion of the cellular Smad4 gene. These results define a novel biochemical property of Smad proteins that is likely to play a direct role in the biologic responses to TGF beta and related ligands.