Merck
  • Evaluation of an iodoform paste in root canal therapy for infected primary teeth.
Evaluation of an iodoform paste in root canal therapy for infected primary teeth.
ASDC journal of dentistry for children (1987-01-01)
F Garcia-Godoy
ABSTRACT
The treatment guidelines for success were if clinically the tooth was painless, without pathologic mobility, and the gingiva was healthy. No clinical or radiographic signs or symptoms of failure were observed in forty-three of forty-five teeth.
Sigma-Aldrich
Iodoform, 99%
Pricing and availability is not currently available.