Merck
  • A novel form of immune signaling revealed by transmission of the inflammatory mediator serotonin between dendritic cells and T cells.
A novel form of immune signaling revealed by transmission of the inflammatory mediator serotonin between dendritic cells and T cells.
Blood (2005-10-15)
Peta J O'Connell, Xiangbin Wang, Matilde Leon-Ponte, Corrie Griffiths, Sandeep C Pingle, Gerard P Ahern
ABSTRACT
Adaptive immunity is triggered at the immune synapse, where peptide-major histocompatibility complexes and costimulatory molecules expressed by dendritic cells (DCs) are physically presented to T cells. Here we describe transmission of the inflammatory monoamine serotonin (5-hydroxytryptamine [5-HT]) between these cells. DCs take up 5-HT from the microenvironment and from activated T cells (that synthesize 5-HT) and this uptake is inhibited by the antidepressant, fluoxetine. Expression of 5-HT transporters (SERTs) is regulated by DC maturation, exposure to microbial stimuli, and physical interactions with T cells. Significantly, 5-HT sequestered by DCs is stored within LAMP-1+ vesicles and subsequently released via Ca2+-dependent exocytosis, which was confirmed by amperometric recordings. In turn, extracellular 5-HT can reduce T-cell levels of cAMP, a modulator of T-cell activation. Thus, through the uptake of 5-HT at sites of inflammation, and from activated T cells, DCs may shuttle 5-HT to naive T cells and thereby modulate T-cell proliferation and differentiation. These data constitute the first direct measurement of triggered exocytosis by DCs and reveal a new and rapid type of signaling that may be optimized by the intimate synaptic environment between DCs and T cells. Moreover, these results highlight an important role for 5-HT signaling in immune function and the potential consequences of commonly used drugs that target 5-HT uptake and release.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, Ī³-irradiated, BioXtra, suitable for cell culture