Merck
  • Direct determination of benzamides in serum by column-switching high-performance liquid chromatography.
Direct determination of benzamides in serum by column-switching high-performance liquid chromatography.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2003-05-29)
Ryoko Chiba, Ayako Ogasawara, Teppei Kubo, Hiroyuki Yamazaki, Masuo Umino, Yoichi Ishizuka
ABSTRACT
A column-switching high-performance liquid chromatographic method with fluorescence detection was developed for the simultaneous determination of four benzamide-type anti-psychotic drugs: sulpiride, tiapride, sultopride and metoclopramide in human serum. In this method, a TSKgel Super-ODS column was used as an analytical column, and a TSKgel G 2000SW was prepared as a pretreatment column. Under the optimized analytical conditions, four benzamide-type anti-psychotic drugs were eluted within 18 min. The detection limits (S/N = 3) for sulpiride, tiapride, sultopride and metoclopramide are 1 ng/ml, 4 ng/ml, 2 ng/ml and 0.5 ng/ml, respectively. Finally, the method was applied to the determination of sulpiride in human serum samples obtained after a single oral dose of sulpiride.
Sigma-Aldrich
Benzamide, 99%
Sigma-Aldrich
Benzamide, purified by sublimation, ≥99.5%
Sigma-Aldrich
Benzamide, zone-refined, purified by sublimation, 99.9%