Merck
  • The role of glutathione S-transferase- and cytochrome P450-dependent metabolism in the olfactory toxicity of methyl iodide in the rat.
The role of glutathione S-transferase- and cytochrome P450-dependent metabolism in the olfactory toxicity of methyl iodide in the rat.
Archives of toxicology (1998-08-26)
M P Chamberlain, E A Lock, B A Gaskell, C J Reed
ABSTRACT
The aim of this study was to investigate the role of metabolic activation in the olfactory toxicity of methyl iodide (MeI). Adult male rats were exposed via nose-only inhalation to 100 ppm MeI for 0-6 h, and non-protein sulphydryl (NP-SH) concentrations determined in selected tissues. Depletion of NP-SH occurred in all tissues, but was most marked and rapid in the respiratory epithelium of the nasal cavity and the kidney. Olfactory, lung and liver NP-SH levels were affected to a lesser extent, and those of the brain declined by only 20-30% over the whole time course. In order to modulate glutathione (GSH) status, animals were pre-treated with (1) phorone plus L-buthionine sulphoximine (BSO), which depleted NP-SH levels in all the tissues examined, or (2) the isopropyl ester of GSH (IP-GSH), which was shown to replenish NP-SH concentrations in all tissues except the liver of animals previously administered phorone. When animals were pre-treated with phorone plus BSO and then exposed to 100 ppm MeI for 2 h, there was a potentiation of the toxicity of MeI as judged by the clinical observations on the animals. In contrast, treatment with IP-GSH prior to and during exposure to MeI for 4 h afforded a marked protection to the olfactory epithelium. In order to inhibit cytochromes P450, animals were pre-treated with cobalt protoporphyrin IX. This decreased hepatic cytochrome P450 concentrations by > 90%, but when animals were then exposed to 100 ppm MeI for 4 h there was no effect on the severity of the olfactory lesion. These results indicate that conjugation of MeI with GSH is a detoxification rather than an activation pathway. Also, there is no major role for cytochrome P450-dependent oxidation in the development of the olfactory lesion.