Merck
  • Cloricromene reduces myocardial infarct size in rabbits when administered during the early reperfusion period.
Cloricromene reduces myocardial infarct size in rabbits when administered during the early reperfusion period.
Anesthesia and analgesia (1997-02-01)
D A Zvara, H J Galaska, V P Castellano, J Vinten-Johansen, R L Royster, M W Williams, B A Murphy, E J Kim
ABSTRACT
Cloricromene is a coumarin derivative without anticoagulant activities that has recently been found to decrease myocardial infarct size after an ischemic-reperfusion injury. This study seeks to determine when the cardioprotective action of cloricromene is exerted in an in vivo rabbit model of ischemic-reperfusion injury. Forty-nine rabbits subjected to 30 min of coronary occlusion and 120 min of reperfusion were randomized into five groups: VEH (n = 11) received saline vehicle; IR (n = 9) received an infusion of cloricromene starting at the onset of ischemia at 8 micrograms.kg-1.min-1; R(-5)(n = 9) and R(+30)(n = 9) received an infusion of cloricromene at 8 micrograms.kg-1.min-1 starting 5 min before reperfusion and 30 min after reperfusion, respectively; and RB(-5)(n = 11) received 300 micrograms/kg bolus of cloricromene 5 min before reperfusion followed by an infusion of 8 micrograms.kg-1.min-1. All infusions were continued until the end of the reperfusion period. Myocardial infarct size was significantly reduced in groups IR, R(-5), and RB(-5). We conclude that cloricromene's effective time of action occurs prior to the first 30 min of the reperfusion period.
Sigma-Aldrich
Cloricromene, ≥98% (HPLC), solid