Merck
  • Formation of farnesal and 3-hydroxy-2,3-dihydrofarnesal from farnesol by protoplasts of Botryococcus braunii.
Formation of farnesal and 3-hydroxy-2,3-dihydrofarnesal from farnesol by protoplasts of Botryococcus braunii.
Biochemical and biophysical research communications (1993-11-15)
H Inoue, T Korenaga, H Sagami, T Koyama, H Sugiyama, K Ogura
ABSTRACT
Farnesal and 3-hydroxy-2,3-dihydrofarnesal (3-hydroxy-3,7,11-trimethyl-6,10-dodecadiene-1-al) were formed from farnesol when the alcohol was incubated with the protoplast of Botryococcus braunii B race strain. This fact suggests the existence of farnesal hydratase in the alga. Feeding experiments showed that both farnesal and 3-hydroxy-2,3-dihydrofarnesal were efficiently incorporated into botryococcenes, triterpenoid hydrocarbons of the alga.
Sigma-Aldrich
Farnesal, mixture of isomers, technical
Sigma-Aldrich
Farnesal, mixture of isomers, ≥85%