- [Mutagenic effect of pindone on D. melanogaster].
[Mutagenic effect of pindone on D. melanogaster].
Bollettino della Societa italiana di biologia sperimentale (1993-04-01)
P Santoro, G Parisi, S Copetti
PMID8129904
ABSTRACT
A commercial grade Pindone was tested for the induction of genetic damage in male germ cells of Drosophila melanogaster by adult ingestion. Adults were starved for 4 days and then fed pindone at a concentration of 1 mM in ethyl alcohol 15% containing sucrose 5%. Pindone was found to increase the frequency of sex chromosome loss. Results were significant at the 1% level (Kastenbaum-Bowman test). However no significant results were obtained by the sex linked recessive lethal assay.
MATERIALS